Learn More
Ampco Flat Back Scratch Brush
SDP
Catalog No. 19173551 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
7 -1/4 in.
Catalog No. Length (English)
19-173-551 7 -1/4 in.
1 options
Catalog No. 19-173-551 Supplier Ampco Supplier No. B401
Add to Cart
Add to Cart
Compact cleaning tool to handle tough residue in harsh or hazardous environments.
  • Independently tested and found to pose no risk in MRI environments using a static magnetic field of 3-Tesla and a maximum spatial gradient magnetic field of 720-Gauss/cm or less.
  • Conforms to ASTM F2503.
  • Meets the requirements established by ASTM A342.
  • Block size: 2 and 1/4 in. x 7 and 1/4 in.
  • Trim length: 1 and 3/4 in.
Specifications
Product Type Flat Back Brush
Length (English) 7 -1/4 in.
Length (English) Bristle 1 -3/4 in.
Quantity 1 Ea.