Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Flat Bottom Boiling Flasks with Interchangeable Joint
SCI_ED

Catalog No. S27051 Shop All Eisco Products
Change view
Click to view available options
Capacity (Metric):
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S27051 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27051 Supplier Eisco™ Supplier No. CH0412I01
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High quality borosilicate 3.3 glass flat bottom boiling flasks feature short necks and interchangeable joints.

  • Flat bottom boiling flask with short neck
  • Made of high quality borosilicate 3.3 glass
  • Ground interchangeable joint
  • Variety of standard socket sizes available
  • Complies with DIN ISO 4797 standard
  • Autoclavable
  • Compliance

    DIN ISO 4797 standard

    Warranty and Services

    1 year limited manufacturer warranty

    Specifications

    Color Clear
    Capacity (Metric) 500 mL
    Material Borosilicate Glass (3.3)
    For Use With (Application) Heating Liquids
    Neck Style Short
    No. of Positions 1
    Product Type Boiling Flask
    Socket Joint 19/26
    Autoclavable Autoclavable
    Disposable Reusable
    Graduated No
    Mouth Style Joint
    Sterility Non-sterile
    Bottom Shape Flat
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.