Learn More
Malvern Panalytical FLAT BOTTOM CRUCIBLE 25ML 30G
Supplier:  Malvern Panalytical P013000
Encompass
Flat Bottom Crucible (Pt-Au5%) 25 ml, 30 G Nominal WeightPt-Au(5%) crucible, volume capacity of 25 ml, weight approximately 30 g.This platinum-gold alloy is universally accepted as the material ofchoice for crucibles and casting molds for XRF analysis. It has highertemperature resistance than pure platinum and has a non-wetting propertythat allows an easy removal of sample after fusion. The crucible hasround internal walls, rim on the outside and a flat bottom.
Catalog No. NC2391967
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.