Learn More
Excelta™ Flat Nose Pliers
Catalog No. 17467392 Shop All Excelta Corporation Products
Change view
Click to view available options
Grade:
Fine Precision
Jaw Type:
Serrated
Smooth
Length (English):
4.75 in.
5.75 in.
6 in.
Tip Style:
Medium Flat Duck-billed Nose
Medium Flat Nose
Small Flat Duck-billed Nose
Small Flat Nose
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade Jaw Type Length (English) Tip Style
17-467-392 Fine Precision Smooth 5.75 in. Medium Flat Duck-billed Nose
17-467-398 Fine Precision Serrated 5.75 in. Medium Flat Nose
17-467-397 Fine Precision Smooth 5.75 in. Medium Flat Nose
17-467-386 Fine Precision Smooth 6 in. Small Flat Duck-billed Nose
17-467-399 Fine Precision Smooth 6 in. Medium Flat Nose
17-456-010 Fine Precision Smooth 4.75 in. Small Flat Nose
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17-467-392 Supplier Excelta™ Supplier No. 2818
Add to Cart
Add to Cart
Have ESD-safe handles with cushioned grip
  • Made of rust-free AISI 440 stainless-steel
Specifications
Grade Fine Precision
Length (English) 5.75 in.
Length (Metric) 14.6 cm
Material AISI 440 Stainless Steel
Grip Type ESD-safe soft foam cushion
Jaw Type Smooth
Tip Style Medium Flat Duck-billed Nose