Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Excelta™ Flat Nose Pliers

Catalog No. 17467399 Shop All Excelta Corporation Products
Change view
Click to view available options
Grade:
Fine Precision
Jaw Type:
Serrated
Smooth
Length (English):
4.75 in.
5.75 in.
6 in.
Tip Style:
Medium Flat Duck-billed Nose
Medium Flat Nose
Small Flat Duck-billed Nose
Small Flat Nose
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade Jaw Type Length (English) Tip Style
17-467-399 Fine Precision Smooth 6 in. Medium Flat Nose
17-467-392 Fine Precision Smooth 5.75 in. Medium Flat Duck-billed Nose
17-467-398 Fine Precision Serrated 5.75 in. Medium Flat Nose
17-467-397 Fine Precision Smooth 5.75 in. Medium Flat Nose
17-467-386 Fine Precision Smooth 6 in. Small Flat Duck-billed Nose
17-456-010 Fine Precision Smooth 4.75 in. Small Flat Nose
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17-467-399 Supplier Excelta™ Supplier No. 2842L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Have ESD-safe handles with cushioned grip

  • Made of rust-free AISI 440 stainless-steel

Specifications

Grade Fine Precision
Length (English) 6 in.
Length (Metric) 15.24 cm
Material AISI 440 Stainless Steel
Grip Type ESD-safe soft foam cushion
Jaw Type Smooth
Tip Style Medium Flat Nose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.