Learn More
3M™ Petrifilm™ Flat Spreader 6425, Sold by AquaPhoenix Scientific
SCI_ED
Catalog No. S30741 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
2 Pk.
Catalog No. Quantity
S30741 2 Pk.
1 options
Catalog No. S30741 Supplier 3M™ Petrifilm™ Supplier No. 6425
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Distributes sample evenly for 3M™ Petrifilm™ Staph Express Count Plates
  • Clear acrylic material allows technician to observe and control spread
  • Spreads inoculum over uniform area
Specifications
Product Type Flat Spreader
For Use With (Equipment) 3M™ Petrifilm™ Staph Express Count Plates
Quantity 2 Pk.