Learn More
Apexbio Technology LLC Thiomyristoyl 1429749-41-6 25mg
SDP
Supplier:  Apexbio Technology LLC B793925
Encompass_Preferred
Thiomyristoyl (CAS 1429749-41-6) is a selective inhibitor of SIRT2 a NAD -dependent deacetylase involved in cell cycle regulation apoptosis metabolic processes and neurodegenerative disease mechanisms By binding specifically to SIRT2 Thiomyristoyl effectively suppresses its enzymatic activity displaying limited activity against other sirtuin family members In vitro studies report an IC50 of approximately 28 nM for SIRT2 inhibition Thiomyristoyl is widely employed as a chemical probe in cellular biology and signal transduction research facilitating investigation of SIRT2-related pathways and supporting the exploration of its biological functions and therapeutic potential
Catalog No. 50-000-91457
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.