Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flavin adenine dinucleotide disodium salt, 100.5%, 95 to 99%, MP Biomedicals™

Catalog No. icn15112780 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15112780 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15112780 Supplier MP Biomedicals, Inc Supplier No. 0215112780
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 146-14-5
Molecular Formula C27H33N9O15P2
Molecular Weight (g/mol) 785.56
MDL Number MFCD00041835
InChI Key VWWQXMAJTJZDQX-UHFFFAOYNA-N
Synonym flavin adenine dinucleotide, flavine-adenine dinucleotide, flavin-adenine dinucleotide, fad, flavitan, fademin, flanin f, unii-zc44yti8kk, flamitajin b, adenine-flavin dinucleotide
PubChem CID 643975
ChEBI CHEBI:16238
IUPAC Name [[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxobenzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl] hydrogen phosphate
SMILES CC1=C(C)C=C2N(CC(O)C(O)C(O)COP(O)(=O)OP(O)(=O)OCC3OC(C(O)C3O)N3C=NC4=C(N)N=CN=C34)C3=NC(=O)NC(=O)C3=NC2=C1

Specifications

Color Orange
Assay Percent Range 95 to 99%
Quantity 100 mg
Formula Weight 829.6
Percent Purity 100.5%
Physical Form Powder
Chemical Name or Material Flavin adenine dinucleotide disodium salt
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.