Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Flavin reductase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580380
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580380 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580380 Supplier Invitrogen™ Supplier No. PA580380
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human BLVRB/biliverdin reductase B.

The final step in heme metabolism in mammals is catalyzed by the cytosolic biliverdin reductase enzymes A and B (EC 1.3.1.24).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Flavin reductase
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene BLVRB
Gene Accession No. P30043
Gene Alias Biliverdin reductase B; biliverdin reductase B (flavin reductase (NADPH)); biliverdin-IX beta-reductase; BLVRB; BVRB; BVR-B; epididymis secretory protein Li 10; flavin reductase (NADPH); FLR; FR; GHBP; green heme-binding protein; HEL-S-10; NADPH; NADPH-dependent diaphorase; NADPH-flavin reductase; SDR43U1; short chain dehydrogenase/reductase family 43U, member 1
Gene Symbols BLVRB
Host Species Rabbit
Immunogen Recombinant Human BLVRB/biliverdin reductase B protein (Ala2-Gln206).
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 645
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.