Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flavone, 99%

Catalog No. AAA1362706
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1362706 5 g
AAA1362703 1 g
AAA1362714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1362706 Supplier Thermo Scientific Chemicals Supplier No. A1362706
Add to Cart

CAS: 525-82-6 | C15H10O2 | 222.243 g/mol

Flavone is a potentially useful biochemical for cytochrome P450 studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Flavone is a potentially useful biochemical for cytochrome P450 studies.

Solubility
Soluble in acetone (25 mg/ml), methanol, alcohol, and chloroform. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers.Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 525-82-6
Molecular Formula C15H10O2
Molecular Weight (g/mol) 222.243
MDL Number MFCD00006825
InChI Key VHBFFQKBGNRLFZ-UHFFFAOYSA-N
Synonym flavone, 2-phenylchromone, 2-phenyl-4h-chromen-4-one, 2-phenyl-4-chromone, 2-phenyl-4-benzopyron, 2-phenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 2-phenyl, asmacoril, chromocor, cromaril
PubChem CID 10680
ChEBI CHEBI:42491
IUPAC Name 2-phenylchromen-4-one
SMILES C1=CC=C(C=C1)C2=CC(=O)C3=CC=CC=C3O2

Specifications

Melting Point 95°C to 98°C
Odor Odorless
Quantity 5 g
Beilstein 157598
Merck Index 14,4092
Solubility Information Soluble in acetone (25mg/ml),methanol,alcohol,and chloroform. Insoluble in water.
Formula Weight 222.24
Percent Purity 99%
Chemical Name or Material Flavone
EINECSNumber 208-383-8
RTECSNumber DJ3100630
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.