Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Flecainide acetate
GREENER_CHOICE

Catalog No. 147010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-701-0 10 mg
14-705-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-701-0 Supplier Tocris Bioscience™ Supplier No. 1470/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cardiac Na+ channel blocker. Antiarrhythmic

Open Na+ channel blocker that inhibits fast Na+ current in cardiac muscle in a use- and concentration-dependent manner. Orally-active class Ic antiarrhythmic agent.

Chemical Identifiers

CAS 54143-56-5
Molecular Formula C19H24F6N2O5

Specifications

Quantity 10 mg
Formula Weight 474.4
Purity >99%
Molecular Weight (g/mol) 474.4
Chemical Name or Material Flecainide acetate
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.