Learn More
Medchemexpress LLC HY-B0414 500mg Medchemexpress, Fleroxacin CAS:79660-72-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0414500MG
Encompass_Preferred
Medchemexpress, HY-B0414 500mg Fleroxacin CAS:79660-72-3 Fleroxacin (RO 23-6240) is a broad-spectrum antimicrobial fluoroquinolone. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2526
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.