Learn More
Market Lab MarketLab Flex-Arm Bioshield
Supplier:  Market Lab 3008
Encompass_Preferred
The Flex-Arm Bioshield is the positionable shield every lab needs. It features al 38"""" reach, stay-put hinges at three key adjustments points (base, elbow and shield), and positioning handles on the shield to help keep the shield clean.
Catalog No. NC9815543
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.