Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOMNI Scientific™ Flexible Tubing
SDP

Catalog No. 04777182 Shop All Somni Scientific Products
Change view
Click to view available options
Color:
Blue
Green
Orange
Red
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
04-777-182 Red
04-777-180 Blue
04-777-183 Orange
04-777-184 Yellow
04-777-185 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 04-777-182 Supplier SOMNI Scientific™ Supplier No. TR2002
Add to Cart
Add to Cart

Provided custom cut per 12-inch increments to your specification. Opaque and firm yet flexible. Colored coded tubing for easy identification.

  • Opaque flexible tubing provided custom cut per 12 inch increments to your specification
  • Tubing is 0.25 in. ID
  • Firm yet flexible PVC
  • Colored coded tubing for easy identification

Specifications

Diameter (English) 0.25 in.
For Use With (Application) Research Animal Inhalant Anesthesia
Color Red
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.