Learn More
Hillrom FlexiPort™ Reusable Blood Pressure Cuff
Description
- FlexiPort technology enables every cuff to be used as a one- or two-tube cuff
- Port can be rotated for improved patient comfort
- Materials do not contain natural rubber latex
- Meet all latest clinical guidelines for proper fit from AAMI and AHA
- Primary sizes are color-coded to make it easy to find the proper cuff size
- Up to 13 sizes, ranging from small infant blood pressure cuffs through large adult blood pressure cuffs (including extra-large, thigh and long cuffs)
Specifications
Specifications
| Size | 10 Small Adult |
| For Use With (Equipment) | Blood Pressure Devices and Patient Monitors |
| Includes | No Tubes or Connectors |
| Latex Free | Yes |
| Product Type | Blood Pressure Cuff |
| Width (Metric) Cuff | 20 to 26 cm |