Learn More
Hillrom FlexiPort™ Reusable Blood Pressure Cuff
Catalog No. 22355159
Change view
Click to view available options
Size:
09 Child
10 Small Adult
11 Adult
12 Large Adult
Includes:
1 Tube, Male Screw Connector
2 Tubes, Tri-Purpose Connector, Inflation Bulb and Valve
No Tubes or Connectors
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Includes
22-355-159 11 Adult 1 Tube, Male Screw Connector
22-355-156 09 Child No Tubes or Connectors
22-355-157 10 Small Adult No Tubes or Connectors
22-355-158 11 Adult No Tubes or Connectors
22-355-160 11 Adult 2 Tubes, Tri-Purpose Connector, Inflation Bulb and Valve
22-355-161 12 Large Adult 2 Tubes, Tri-Purpose Connector, Inflation Bulb and Valve
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 22-355-159 Supplier Hillrom Supplier No. WEL REUSE111SC
Add to Cart
Add to Cart
This item is not returnable. View return policy
Invest in an easy-to-use and affordable reusable blood pressure cuff solution. The single-point FlexiPort snap connection was designed to simplify connectivity, reducing the hassle of needing multiple cuff tubes and connectors. Easily size cuffs up or down for each patient.
  • FlexiPort technology enables every cuff to be used as a one- or two-tube cuff
  • Port can be rotated for improved patient comfort
  • Materials do not contain natural rubber latex
  • Meet all latest clinical guidelines for proper fit from AAMI and AHA
  • Primary sizes are color-coded to make it easy to find the proper cuff size
  • Up to 13 sizes, ranging from small infant blood pressure cuffs through large adult blood pressure cuffs (including extra-large, thigh and long cuffs)
Specifications
Size 11 Adult
For Use With (Equipment) Blood Pressure Devices and Patient Monitors
Includes 1 Tube, Male Screw Connector
Latex Free Yes
Product Type Blood Pressure Cuff
Width (Metric) Cuff 25 to 34 cm