Learn More
Wells Lamont™ FlexTech™ Cut Resistant Gloves with Impact Protection and Nitrile Coating
Shop All Wells Lamont Industry Group ProductsDescription
- Excellent for applications requiring multi-hazard protection, grip, and dexterity
- Machine knit cut resistant shell maximizes protection without sacrificing comfort
- Soft and flexible TPR pads protect the back of the hand and fingers from impacts and blows
- Padded palm features extra protection
- Reinforced thumb crotch extends wear
- Sandy nitrile palm provides outstanding grip in wet and oily conditions
- ANSI Impact Level 1
- ANSI Abrasion Level 5
- ANSI Cut Level A7
- Sold by the dozen pair
Specifications
Specifications
| Construction | Seamless Knit |
| Coating Material | Nitrile |
| Cut Resistance Rating | ANSI A7 |
| Hand | Specific |
| Product Type | Cut-resistant Gloves |
| Size | Medium |
| For Use With (Application) | For applications requiring multi-hazard protection, grip, and dexterity. |
| Glove Cuff Style | Elasticated |