Learn More
Wells Lamont™ FlexTech™ Cut Resistant Gloves with Impact Protection and Nitrile Coating
Catalog No. 19086624 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-086-624 2X-Large
19-086-620 Small
19-086-621 Medium
19-086-622 Large
19-086-623 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-086-624 Supplier Wells Lamont™ Supplier No. I2459XXL
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
FlexTech™ Cut Resistant Gloves with Impact Protection and Nitrile Coating offers an ANSI A7 level of cut resistance along with impact protection for the back of the hand. A sandy nitrile palm coating provides excellent grip in dry, wet, or oily conditions.
  • Excellent for applications requiring multi-hazard protection, grip, and dexterity
  • Machine knit cut resistant shell maximizes protection without sacrificing comfort
  • Soft and flexible TPR pads protect the back of the hand and fingers from impacts and blows
  • Padded palm features extra protection
  • Reinforced thumb crotch extends wear
  • Sandy nitrile palm provides outstanding grip in wet and oily conditions
  • ANSI Impact Level 1
  • ANSI Abrasion Level 5
  • ANSI Cut Level A7
  • Sold by the dozen pair
Specifications
Construction Seamless Knit
Coating Material Nitrile
Cut Resistance Rating ANSI A7
Hand Specific
Product Type Cut-resistant Gloves
Size 2X-Large
For Use With (Application) For applications requiring multi-hazard protection, grip, and dexterity.
Glove Cuff Style Elasticated