Learn More
Wells Lamont™ FlexTech™ Cut Resistant Gloves with Nitrile Coating
Catalog No. 19086708 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-086-708 Large
19-086-706 Small
19-086-707 Medium
19-086-709 X-Large
19-086-710 2X-Large
19-086-705 X-Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-086-708 Supplier Wells Lamont™ Supplier No. Y9216L
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
FlexTech™ Cut Resistant Gloves with Nitrile Coating offer ANSI A7 cut resistance, dexterity, and an excellent grip, especially in wet and oily conditions.
  • Seamless machine knit cut resistant shell provides all-day comfort and protection
  • Sandy nitrile palm coating delivers improved grip in wet and oily applications
  • Machine washable for longer service life
  • 13 Gauge
  • ANSI Abrasion Level 5
  • ANSI Puncture Level 5
  • ANSI Cut Level A7
  • Sold by the dozen pair
Specifications
Construction Seamless Knit
Coating Material Nitrile
Cut Resistance Rating ANSI A7
Hand Specific
Product Type Cut-resistant Gloves
Size Large
For Use With (Application) For improved grip in wet and oily applications.
Glove Cuff Style Elasticated
Model Y9216
Yarn Gauge 13