Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ FlexTech™ Foam Nitrile Coated Nylon Gloves

Catalog No. 19148711 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-148-711 Medium
19-148-709 X-Small
19-148-710 Small
19-148-712 Large
19-148-713 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-148-711 Supplier Wells Lamont™ Supplier No. Y9259M
This item is not returnable. View return policy

Foam nitrile coating provides abrasion resistance and enhanced grip in dry or slightly oily applications.

  • Lightweight synthetic knit shell increases flexibility and comfort
  • Foam nitrile palm enhances grip and resistance to snags & abrasions
  • Ideal for electronics assembly and small parts handling

Specifications

Coated Area Palm
Coating Material Nitrile Foam
Color Gray, White
Hand Specific
Material Nylon
Product Type Dip-coated Gloves
Size Medium
For Use With (Application) Electronics Assembly, and Small Parts Handling
Glove Cuff Style Knit
Series Y9259
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.