Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ FlexTech™ DuPont™ Aramid Fiber Gloves with Foam Nitrile Coating

Catalog No. 19048917 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
2X-Large
Large
Medium
Small
X-Large
X-Small
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-048-917 Large
19-048-914 X-Small
19-048-915 Small
19-048-916 Medium
19-048-918 X-Large
19-048-919 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-048-917 Supplier Wells Lamont™ Supplier No. Y9256L
Add to Cart
Add to Cart
This item is not returnable. View return policy

Foam Nitrile Coated FlexTech™ DuPont™ Aramid Fiber Glove Shells are ideal for a variety of applications that require a high level of dexterity along with cut protection.

  • Lightweight stretch Aramid Fiber provides flexibility and cut protection
  • Foam nitrile palm enhances grip and resistance to snags and abrasions
  • Uncoated back circulates air for breathability to keep hands cool and comfortable
  • Non-slip surface increases efficiency and worker confidence

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance ASTM 1790-97, ASTM F2992-15, ASTM D3389-05, ASTM F1358-08
Coating Material Nitrile Foam
Color Yellow
Cut Resistance Rating ANSI A2
Hand Specific
Material Aramid Fiber
Product Type Cut-resistant Gloves
Size Large
EN388 Standard EN 388:3532
For Use With (Application) General assembly, auto assembly, construction, glass handling, metal fabrication
Glove Cuff Color Brown
Glove Cuff Style Knit
Model Y9256L
Series FlexTech™ Y9256
Yarn Gauge 15
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.