Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ FlexTech™ Polyurethane Coated Seamless Knit HPPE Gloves

Catalog No. 19148717 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-148-717 Large
19-148-714 X-Small
19-148-715 Small
19-148-716 Medium
19-148-718 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-148-717 Supplier Wells Lamont™ Supplier No. Y9265L
Add to Cart
Add to Cart
This item is not returnable. View return policy

FlexTech™ Polyurethane Coated Seamless Knit HPPE Gloves are constructed with High Performance Polyethylene (HPPE) Fibers for increased cut protection. Polyurethane coating on the palm provides a non-slip surface for enhanced grip capabilities.

  • High performance polyethylene fibers (HPPE) blended with 13-gauge nylon for increased cut resistance
  • Gloves combine comfort, dexterity, and cut resistance
  • Polyurethane coating on the palm enhances grip and tactility
  • Launderable for an extended useful life

Specifications

ANSI Abrasion Rating ANSI 6
Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A2
Exterior Finish No Texture
Hand Specific
Material HPPE, Nylon
Product Type Seamless Knit Gloves
Size Large
For Use With (Application) General Assembly
Glove Cuff Style Knit
Series Y9265
Yarn Gauge 13
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

WARNING: Cancer – www.P65Warnings.ca.gov

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.