Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ FlexTech™ Y9219 Polyurethane-Coated Cut-Resistant Gloves

Catalog No. 17200415 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-200-415 X-Large
17-200-412 Small
17-200-413 Medium
17-200-414 Large
17-200-416 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-200-415 Supplier Wells Lamont™ Supplier No. Y9219XL
Add to Cart
Add to Cart

Ultra-lightweight liner with polyurethane coating on the palm delivers dexterity, tactility and comfort. FlexTech™ Y9219 Polyurethane-Coated Cut-Resistant Gloves provide ANSI Cut Level A9 cut resistance for exceptional protection.

  • 18-gauge ultra lightweight liner delivers dexterity, tactility and comfort
  • Constructed from a blend of tungsten, fiberglass, HPPE and polyester
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Compatible with touchscreen devices for easy use with tablets and phones
  • Machine washable for extended wear life
  • ANSI A9 Cut Resistance
  • ANSI Puncture Level 4

Specifications

ANSI Abrasion Rating ANSI 4
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A9
Hand Specific
Material HPPE, Polyester, Tungsten, Fiberglass
Product Type Cut-resistant Gloves
Size X-Large
For Use With (Application) Assembly, metal handling, fabrication and stamping
Glove Cuff Style Knit
Model Y9219XL
Puncture Rating ANSI/ISEA 105-16 Level 4
Series FlexTech™ Y9219
Yarn Gauge 18
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.