Learn More
Wells Lamont™ FlexTech™ Y9298 Cut Resistant Gloves
Catalog No. 17000490 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-000-490 X-Large
17-000-487 Small
17-000-488 Medium
17-000-489 Large
17-000-491 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-000-490 Supplier Wells Lamont™ Supplier No. Y9298XL
Add to Cart
Add to Cart
The FlexTech™ Y9298 Cut Resistant Gloves combine an ultra lightweight liner that provides excellent dexterity with an ANSI Cut Level A4 protection to reduce hand injuries.
  • High-cut resistant yarn provides optimal cut protection to reduce hand injuries
  • 18 gauge ultra light weight liner delivers ultimate dexterity and comfort
  • Composite yarn design allows increased dexterity during process operations
  • Touchscreen compatible for easy use with tablets and phones
  • Polyurethane coated for enhanced abrasion resistance and grip
  • Machine washable for longevity
Specifications
Construction Knit
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A4
Hand Specific
Latex Free No
Material Stainless Steel Fiber, Fiberglass, Nylon, HPPE
Product Type Cut-resistant Gloves
Size X-Large
Gender Unisex
Glove Cuff Style Elasticated
Puncture Rating ANSI/ISEA 105-16 Level 4
Thumb Style Straight
Yarn Gauge 18
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov