Learn More
AdipoGen Flibanserin
SDP
Supplier:  AdipoGen CDXF0337M025
Encompass_Preferred
Chemical. CAS 167933-07-5. Formula C20H21F3N4O. MW 390.4. Synthetic. Flibanserin is a full agonist of the serotonin 5-HT1A receptor and an antagonist of 5-HT2A Ki=1nm and 49nM, respectively. It also binds to dopamine D4 receptors with Ki values ranging from 4-24nM, but demonstrates no affinity for the other 5-HT subtypes or other neurotransmitter receptors. Multifunctional serotonergic ligands like flibanserin that can enhance downstream release of dopamine and norepinephrine while concomitantly reducing serotonin release in the brain circuits that mediate symptoms of reduced sexual interest and desire, are being studied clinically for therapeutic potential to improve sexual functioning. Flibanserin was investigated as a novel, non-hormonal treatment for pre-menopausal women with Hypoactive Sexual Desire Disorder HSDD. Flibanserin was also found to reduce L-DOPA-induced dyskinesia in a model of Parkinson‘s Disease.
Catalog No. 50-256-0854
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.