Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fischer Technical Company™ Flip Style Two Valve Natural Rubber Safety Pipet Filler
SCI_ED

Catalog No. S012383 Shop All Fischer Technical Company Products
Change view
Click to view available options
Color:
Black
Blue
Green
Red
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S012383 Yellow
S012385 Blue
S012384 Green
S32247 Red
S012386 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Prop 65: Cancer and Reproductive Harm.
Catalog No. S012383 Supplier Fischer Technical Company™ Supplier No. FLIP005
Add to Cart
Add to Cart
Prop 65: Cancer and Reproductive Harm.

Safety Pipet Filler is easy to use and clean.

This patented ball fits pipets up to 50 mL.

  • Press on the bulb to evacuate
  • Press on the up arrow to intake solution
  • Drain by pressing on the down arrow
  • Top valve can be removed and reinserted for easy cleaning of the inside

Specifications

Autoclavable Autoclavable
Color Yellow
Disposable Reusable
For Use With Pipette
Grip Type Round
Material Natural Rubber
Model Flip
Number of Channels 1
Sterility Non-sterile
Type Pipette Filler
Volume (Metric) 50 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.