Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ FLIPR™ Liberty Robotic Tips
GREENER_CHOICE

Catalog No. 14222139 Shop All Axygen Products
Change view
Click to view available options
Color:
Black
Clear
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-222-139 Black
14-222-140 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-222-139 Supplier Axygen™ Supplier No. LT384BKR
Add to Cart
Add to Cart
Request a Quote

Accurate sample delivery

  • Available in clear and black
  • Volume: 30μL
  • Available as racked or racked and pre-sterilized
  • For use with Molecular Devices Liberty head
  • Packaging: 384 Tips/rack; 10 Racks/unit; 5 Units/Cs.

Certifications

Certified RNase-, DNase- and endotoxin-safe

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 30 μL
Sterility Non-sterile
Color Black
Autoclavable Autoclavable
Clean Claims DNase, RNase, and Pyrogen free
Format Rack
Tip Style Automation, Round
Packaging SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) Molecular Devices FLIPR 384-well Head
Product Line FLIPR
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.