Learn More
Thermo Scientific™ FlipStrip™ Microtube Racks with Lid
GREENER_CHOICE
Catalog No. 2140236
Change view
Click to view available options
Color:
Fluorescent Green
Fluorescent Orange
Fluorescent Pink
Red
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-402-36 Fluorescent Orange
21-402-35 Fluorescent Pink
21-402-37 Fluorescent Green
21-402-34 Yellow
21-402-38 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 21-402-36 Supplier Thermo Scientific™ Supplier No. 8678
Add to Cart
Add to Cart
Hold 0.2mL strips and 1.5mL tubes on one side and and 0.5mL tubes on the other side of this microtube rack with lid, which is made from 100% recycled plastic.
Hold 0.2mL strips and 1.5mL tubes on one side of the Thermo Scientific™ FlipStrip Microtube Rack with Lid, and 0.5mL tubes on the other. The rack is made from 100% recycled plastic.
  • Holds 0.2mL strips and 1.5mL tubes on one side, 0.5mL tubes on other
  • Stable, low-profile design with an alphanumeric grid for easy tube identification
  • Lid protects samples and allows racks to be stacked
  • Several colors available
TRUSTED_SUSTAINABILITY
Specifications
Description Flipstrip Rack
Autoclavable Not Autoclavable
Color Fluorescent Orange
Holds Microtubes (0.5 mL), Microtubes (1.5 mL), Microtubes (2 mL)
Material Recycled Plastic
Shape Rectangular
Stackable Yes
No. per Case 12 Racks
Product Type Flipstrip Rack