Learn More
Celestron™ Flipview 5MP LCD Portable Microscope
SCI_ED
Catalog No. S43584 Shop All Celestron International Products
Change view
Click to view available options
Magnification Power:
10X to 300X (1200X with 4X digital zoom)
Catalog No. Magnification Power
S43584 10X to 300X (1200X with 4X digital zoom)
1 options
Catalog No. S43584 Supplier Celestron™ Supplier No. 44314
5MP Handheld Portable Digital Microscope with integrated LCD screen.
  • Perfect for up-close observation
  • Onboard 3 in. full color TFT LCD screen flips out when needed, then folds back into microscope body for portability
  • Rechargeable lithium ion battery provides hours of in-the-field viewing
  • FlipView, with 10X to 120X magnification is designed for viewing an array of 3D specimens
FlipView can be used in three configurations:
  • Use it in handheld mode for quick and easy observations
  • Flip out the attached leg: for viewing flat objects like stamps, currency
  • For longer observing sessions of 3D objects, mount FlipView to the included metal stand via the ¼-20 connector. The stand features two articulation points and can be adjusted to point it precisely at subject
Specifications
Magnification Power 10X to 300X (1200X with 4X digital zoom)