Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLJ11506 Antibody [FITC], R&D Systems™
SDP

Catalog No. NBP318422F
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP318422F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP318422F Supplier R&D Systems Supplier No. NBP318422F

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

FLJ11506 Polyclonal antibody specifically detects FLJ11506 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot

Specifications

Antigen FLJ11506
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias AAGAB, alpha- and gamma-adaptin-binding protein p34, KPPP1, p34, PPKP1A
Host Species Rabbit
Immunogen Recognizes region between residues Between Between 265 and 315..
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 79719
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.