Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ FLJ22624 Recombinant Protein

Catalog No. 89013540 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-013-540 10 μg
89-013-541 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-013-540 Supplier Abnova™ Supplier No. H00079866P0110
Add to Cart
Add to Cart

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Accession Number AAH56876
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 79866
Molecular Weight (g/mol) 86.35
Name FLJ22624 (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Source Wheat Germ (in vitro)
Immunogen MQITPETPGRIPVLNPFESPSDYSNLHEQTLASPSVFKSTKLPTPGKFRWSIDQLAVINPVEIDPEDIHRQALYLSHSRIDKDVEDKRQKAIEEFFTKDVIVPSPWTDHEGKQLSQCHSSKCTNINSDSPVGKKLTIHSEKSDAACQTLLSLPVDFNLENILGDYFRADEFADQSPGNLSSSSLRRKLFLDGNGSISDSLPSASPGSPHSGVQTSLEMFYSIDLSPVKCRSPLQTPSSGQFSSSPIQASAKKYSLGSITSPSPISSPTFSPIEFQIGETPLSEQRKFTVHSPDASSGTNSNGITNPCIRSPYIDGCSPIKNWSPMRLQMYSGGTQYRTSVIQIPFTLETQGEDEEDKENIPSTDVSSPAMDAAGIHLRQFSNEASTHGTHLVVTAMSVTQNQSSASEKELALLQDVEREKDNNTVDMVDPIEIADETTWIKEPVDNGSLPMTDFVSGIAFSIENSHMCMSPLAESSVIPCESSNIQMDSGYNTQNCGSNIMDTVGAESYCKESDAQTCEVESKSQAFNMKQDHTTQRCWMKTASPFQCSSP
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias BORA/FLJ22624
Common Name C13orf34
Gene Symbol C13orf34
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.