Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLK-1 Rat anti-Mouse, APC, Clone: Avas 12alpha1, BD

Catalog No. p-7092721 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
25 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560070 0.1 mg
BDB555307 0.5 mg
BDB561993 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BDB560070 Supplier BD Biosciences Supplier No. 560070
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The Avas 12α1 monoclonal antibody specifically binds to fetal liver kinase 1 (Flk-1), a receptor protein tyrosine kinase closely related to CD117 (c-kit) and CD140a (PDGF Receptor α chain) of the immunoglobulin superfamily. Flk-1, also known as VEGF Receptor-2 (VEGF-R2), is a receptor for vascular endothelial growth factor (VEGF). It is expressed, at the mRNA and protein levels, on distinct sets of mesoderm during gastrulation and on endothelial cells in embryonic and adult tissues. In vivo and in vitro studies indicate that Flk-1 is required for the embryonic development of vascular endothelial and hematopoietic cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen FLK-1
Applications Flow Cytometry
Classification Monoclonal
Clone Avas 12alpha1
Concentration 0.2mg/mL
Conjugate APC
Description Fetal liver kinase 1; CD309; Kdr; VEGF receptor-2; VEGFR-2
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse FLK-1 Recombinant Protein
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.