Learn More
Foxx Life Sciences EZBiopure Flask Assembly, 500mL, 38-430 mm Cap, PolyCarbonate (PC) Flat Base Flask with, EZBioWeld Tubing and AseptiQuik S, 8/CS
Supplier:  Foxx Life Sciences 5689733FLS
Encompass_Preferred
The EZBio Single Use Sterile Erlenmeyer Flask Assembly with TPE Tubing and Vent Filter is designed for fluid transfer and storage. Available in polycarbonate (PC) flask Assemblies or Cap assembly, made with USP Class VI components and Gamma Irradiated for sterility and quality
The EZBio MTO Flask Assemblies allow customized component selection to suit project needs. Customers can choose the bottle volume, male or female connectors, and TPE tubing brand.
Features
Ideal for single use, replacing stainless-steel equipment.
Available in baffled and flat base options with volumes of 1/2L.
Sterility tested to SAL 10-6. Both the flask and VersaCap are autoclavable.
Application
Cell culture incubation
Seed flask inoculation
Bioreactor fed-batch/additions
Media and buffer transfer
Bioprocess Product sampling and supplement injection
Product storage
Filtration and purification process connection
Cell harvesting
Catalog No. 50-255-8932
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.