Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform™ Floor Marking Tape

Catalog No. 19169775 Shop All AccuformNMC Products
Change view
Click to view available options
Color:
Blue
Green
Red
Yellow
Dimensions (L x W):
108 ft. x 2 in.
108 ft. x 3 in.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Dimensions (L x W)
19-169-775 Blue 108 ft. x 3 in.
19-169-774 Blue 108 ft. x 2 in.
19-169-776 Green 108 ft. x 3 in.
19-169-777 Red 108 ft. x 3 in.
19-169-778 Yellow 108 ft. x 3 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-169-775 Supplier Accuform™ Supplier No. PTM633BU
Add to Cart
Add to Cart

Accuform™ Floor Marking Tape is used to mark specific areas and promote safety by identifying potential hazard areas

  • Vinyl adheres to floors, walls, piping, posts, beams, handrails, stairways, machinery, and equipment
  • Waterproof, and wear-resistant
  • Minor resistance to acids, oils, grease, caustics, alkalis, and most solvents
  • Permanent self-adhesive back with no liner

Specifications

Color Blue
Dimensions (L x W) 108 ft. x 3 in.
Thickness 5 mil
Material Adhesive Vinyl
Type Marking Tape
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.