Learn More
Accuform™ Floor Sign - THANK YOU FOR PRACTICING SOCIAL DISTANCING KEEP 6 FT FROM PERSON IN FRONT
Catalog No. 19169663 Shop All AccuformNMC Products
Change view
Click to view available options
Color:
Black on Orange
Black on Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-169-663 -
19-169-666 Black on Yellow
19-169-667 -
19-169-665 Black on Orange
19-169-664 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-169-663 Supplier Accuform™ Supplier No. PSR300
Accuform™ Floor Signs alert and remind everyone to practice social and physical distancing to prevent the spread of the Coronavirus (COVID-19)
  • Constructed of thick adhesive vinyl
  • Subsurface printed and laminated to protect the graphics and message from heavy pedestrian traffic
  • Non-slip, pebble-textured surface provides added durability
  • Strong, permanent, acrylic adhesive back and peel-off liner
  • Adheres firmly to smooth, clean, dry surfaces
Specifications
Mounting Type Self-adhesive
Languages English
Dimensions (L x W) 12 x 18 in.