Learn More
Accuform™ Floor Sign - THANK YOU FOR WAITING IN LINE UNTIL YOU ARE CALLED UPON
Catalog No. 19169640 Shop All AccuformNMC Products
Change view
Click to view available options
Dimensions (L x W):
12 x 12 in.
17 x 17 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-169-640 12 x 12 in.
19-169-641 17 x 17 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-169-640 Supplier Accuform™ Supplier No. MFS341
Add to Cart
Add to Cart
Accuform™ Floor Signs alert and remind everyone to practice social and physical distancing to prevent the spread of the Coronavirus (COVID-19)
  • Constructed of thick adhesive vinyl
  • Subsurface printed and laminated to protect the graphics and message from heavy pedestrian traffic
  • Non-slip, pebble-textured surface provides added durability
  • Strong, permanent, acrylic adhesive back and peel-off liner
  • Adheres firmly to smooth, clean, dry surfaces
Specifications
Mounting Type Self-adhesive
Languages English
Dimensions (L x W) 12 x 12 in.
Displayed Text THANK YOU FOR WAITING IN LINE UNTIL YOU ARE CALLED UPON
Color Black on Yellow