Learn More
Adam Equipment GFK 1320A Floor weighing scale, 1320lb / 600kg Capacity and 0.1lb / 0.05kg Readability
Supplier: Adam Equipment GFK 1320A
Adam Equipment GFK Bench Checkweighing Scales feature a full keypad that simplifies unit weight and sample entry, the GFK-M floor scale capably performs checkcounting to a predefined weight or count. Ideal for manufacturing, food processing, stock management and inventory control procedures in warehouse or factory environments. A durable stainless steel platform and pillar-mounted ABS plastic indicator make the GFK suitable for most counting and weighing applications. The large backlit display is visible in most settings and shows unit weight, total weight and number of pieces. GFK-M models are NTEP trade certified for selling products by weight. With large platforms, high capacities and grade 304 stainless steel pans, GFK and GFK-M scales are ideal for demanding industrial use, including food processing and manufacturing tasks. The GFK-M version is approved for selling by weight. Rechargeable battery included for operation almost anywhere.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.