Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Flow adapter for Luer lock LC Columns, Column I.D. 2.5 cm

Catalog No. 111009898 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Diameter (Metric) Inner:
2.5 cm Column ID
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner
11-100-9898 2.5 cm Column ID
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-9898 Supplier MilliporeSigma™ Supelco™ Supplier No. F85171EA
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Flow adapters consist of 1/16 in. O.D. FEP tubing, a polyacetal body, a Viton™ O-ring seal and a 20 μm porosity HDPE bed support.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Inner 2.5 cm Column ID
For Use With (Application) Chromatography Applications

Viton is a registered trademark of E. I. du Pont de Nemours and Company

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.