Learn More
Beckman Coulter Flow Check Fluorospheres
Supplier:  Beckman Coulter 6605359
Encompass
Flow-Check Fluorospheres consist of 10 um (nominal diameter) polystyrene fluorescent microspheres suspended in an aqueous medium containing surfactants and preservatives at 1 x 106 fluorospheres/mL (nominal concentration). The fluorescence emission of the dye contained within the fluorospheres ranges from 525 nm to 700 nm when excited at 488 nm. Flow-Check Fluorospheres are used to verify instrument optical alignment and fluidics.
Catalog No. NC1945482
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.