Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Flow Control Adapter
GREENER_CHOICESDP

Catalog No. 31501004 Shop All Kemtech Products
Change view
Click to view available options
Material:
Borosilicate Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
31-501-004 Borosilicate Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 31-501-004 Supplier Synthware™ Supplier No. C432440
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Features a 0 to 4 mm high vacuum PTFE valve with metering tip to adjust the air pressure.

O-rings on the valve are completely encapsulated with a thin layer of PTFE, making it inert to most chemicals. For use with columns having a Rodaviss 24/40 outer joint.

Specifications

Type Adapter
Color Clear
For Use With (Application) Organic Synthesis
Material Borosilicate Glass
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.