Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Flow Cytometry Permeabilization/Wash Buffer I (1X)
SDP

Catalog No. FC005NOV Shop All Novus Products
Change view
Click to view available options
Quantity:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FC005NOV 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FC005NOV Supplier Novus Biologicals™ Supplier No. FC005NOV

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Formulated and optimized for permeabilization of single-cell suspensions, dilution and washing of these samples for subsequent intracellular flow cytometry applications

Specifications

Product Type Permeabilization/Wash Buffer
Content And Storage Store at 4°C. Do not freeze.
Quantity 250mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.