Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Flow Reducing Washer

Catalog No. XX1004710 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
300 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
XX1004710 300 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. XX1004710 Supplier MilliporeSigma™ Supplier No. XX1004710
Add to Cart
Add to Cart

Specifications

For Use With (Equipment) 47 mm Glass Filter Holders
No. per Pack 300Pack
Quantity 300 Pack
Type Flow-reducing washer, Whole
Diameter (Metric) Inside 25 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.