Learn More
Bel-Art™ Flowmi™ Cell Strainers for 1000μL Pipette Tips
Shop All SP Bel-Art & SP Wilmad-LabGlass Products

Description
- Provide fast and efficient filtering of small volume samples (up to 1000 μL) prior to FLOW or FACS analysis
- Preserves volume when filtering small sample volumes
- Removes cell aggregates and creates uniform single cell suspensions
- Decreases the potential for clogging of FLOW or FACS instruments by effectively filtering cellular debris
- Compact tray holds 50 Flowmi Cell Strainers ready for press and fit attachment to tip; features a sliding cover for easy, one-handed use
- Packaged in a re-sealable bag to maintain sterility when used in laminar flow hood or biohazard hood
- Sterile, 40 μm or 70 μm porosity
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Mesh Material | Polyethylene |
| Color | White |
| For Use With (Application) | Provide fast and efficient filtering of small volume samples (up to 1000μL) prior to FLOW or FACS™ analysis |
| Includes | Compact tray |
| Product Type | Cell Strainer |
| Sterility | Sterile |
| For Use With (Equipment) | Pipette Tips, 1000 μL, including Fisherbrand Sure-One, VWR, Axygen, Nichiryo, Eppendorf and more |
| Mesh Size | 70 μm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.