Learn More
Electron Microscopy Sciences FlowView Starter Kit - Bio
SDP
Supplier:  Electron Microscopy Sciences FV104
Encompass_Preferred
FlowView starter kit provides world-leading innovative liquid sample inspection with SEM. These economic and easy-to-use starter kits include stage, accessories, and 24 Microscopic Fluid Chips. The chips are compatible with SEM for most major brands. Kit contains tweezer, pipette, torque wrench, carrier box, storage box, and 24 microscopic fluid chips. Use in cell morphology, bio-fluidic microchips, and polystyrene. Tweezer, pipette, torque wrench, carrier box, storage box, 24 microscopic fluid chips.
Catalog No. 50-192-8104
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.