Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ FlowX FoxP3/Transcription Factor Fixation & Perm Buffer Kit
SDP

Catalog No. FC012100
Change view
Click to view available options
Product Type:
Fixative
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
FC012100 Fixative
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FC012100 Supplier R&D Systems™ Supplier No. FC012100

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Applications: Flow Cytometry

FlowX™ FoxP3/Transcription Factor Fixation & Permeabilization Buffer Kit is formulated and optimized for immunofluorescent staining of FoxP3+ or other transcription factor positive single-cell suspensions intended for flow cytometric acquisition and analysis. This product is supplemented with the metabolic inhibitor sodium azide.

Specifications

Content And Storage Store the unopened product at 2 - 8° C. Do not use past expiration date.
Product Type Fixative
Quantity 100 Tests
Formulation Supplied in in a buffered protein base with preservatives.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.