Learn More
R&D Systems™ FlowX Human Th17 Cell Multi-Color Flow Cytometry Kit
Description
- This kit contains five conjugated antibodies and buffers that can be used for staining of human Th17 cells (1-12)
- Positive Markers:
- 125μL of CD4 Alexa Fluor™ 594 Mouse IgG2A (Clone 11830)
- 125μL of IL-17A Alexa Fluor™ 700 Mouse IgG2B (Clone 41809)
- 250μL of RORα-PE Mouse IgG2B (Clone 784652)
- 125μL of RORγt Alexa Fluor™ 488 Rabbit IgG (Clone 1181A)
- 250μL of CXCR6 APC Mouse IgG2B (Clone 56811)
- FoxP3/Transcription Factor Fixation Concentrate (4X):
- 8mL of a formaldehyde solution.
- FoxP3/Transcription Factor Fixation Diluent:
- 25mL of a buffered detergent
- FoxP3/Transcription Factor Permeabilization and Wash Buffer (10X)
- 25mL of a buffered protein base with detergent and preservatives. May contain a precipitate but will not affect product performance.
- Staining Buffer (1X):
- 50mL of a 1X staining buffer
- Some components in this kit contain formaldehyde which is a suspected carcinogen.
- Avoid contact with skin, eyes, and mucous membranes, and avoid inhaling fumes.
- In case of contact, wash immediately with water and seek medical advice.
- Some components in this kit contain sodium azide which may react with lead and copper plumbing to form explosive metallic azides.
- Flush with large volumes of water during disposal.
Specifications
Specifications
| Quantity | 25 Tests |
| No. of Tests | 25 |
| Content And Storage | Store the unopened product at 2 - 8° C. Do not use past expiration date. |