Learn More
R&D Systems™ FlowX Rat Regulatory T Cell Multi-Color Flow Kit
Description
- This kit contains three conjugated antibodies (and a mouse IgG1 isotype control) that can be used for single-step staining of rat regulatory Treg cells (1-9).
- Positive Markers:
- 125μL of FoxP3 Alexa Fluor™ 647 Mouse IgG1 (Clone 376209)
- 250μL of CD25/IL-2 Rα-PE Mouse IgG1 (Clone 745520)
- 250μL of CD4-Fluorescein Mouse IgG2A (Clone OX-38)
- Isotype Control:
- 250μL of Mouse IgG1 Alexa Fluor™ 647 Mouse IgG1 (Clone 11711)
- FoxP3/Transcription Factor Fixation Concentrate (4X):
- 8mL of a formaldehyde solution.
- FoxP3/Transcription Factor Fixation Diluent:
- 25mL of a buffered detergent.
- FoxP3/Transcription Factor Permeabilization & Wash Buffer (10X):
- 25mL of a buffered protein base with detergent and preservatives. May contain a precipitate but will not affect product performance.
- Staining Buffer (1X):
- 50mL of a 1X staining buffer.
- Some components in this kit contain formaldehyde which is a suspected carcinogen.
- Avoid contact with skin, eyes, and mucous membranes, and avoid inhaling fumes.
- In case of contact, wash immediately with water and seek medical advice.
- Some components in this kit contain sodium azide which may react with lead and copper plumbing to form explosive metallic azides.
- Flush with large volumes of water during disposal.
Specifications
Specifications
| Quantity | 25 Tests |
| No. of Tests | 25 |
| Content And Storage | Store the unopened product at 2 - 8° C. Do not use past expiration date. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.