Learn More
TRANE Technologies FARRAR Lab Grade Upright Refrigerator +4°C (FLR125)
Supplier:  TRANE Technologies 5118125FAR115
Encompass_Preferred
FARRAR Lab Grade Refrigerators are ENERGY STAR certified and SNAP/EU F-gas-compliant, pairing a powerful variable capacity compressor with a low global warming potential (GWP) natural hydrocarbon refrigerant (R600a). These refrigerators are suitable for lab environments with ultra-quiet operation (42dB – 52dB).With a temperature range of +2°C to +10°C with +/-1°C uniformity, FARRAR refrigerators offer multiple sizes in single or double door configurations with premium features like glass doors, interior lights, and a high reliability, long life solid buffer thermal ballast. These refrigerators have undergone detailed Accelerated Life Testing (ALT) to support a 10-year service life, providing high reliability and performance.Specifications for this model (FLR125)* Capacity - 25.2 cu ft (714 L)* Shelves/Drawers - 4 adjustable shelves and locking drawer available as accessory* Exterior Dimensions - 29 x 78.9 x 34.1 in* Electrical - dual voltage: 115V 60Hz or 220-240V 50/60Hz
Catalog No. NC3257200
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.