Learn More
Wilmad Labglass FLSK VOL CL A SRLZD 10ML
Supplier:  Wilmad Labglass LG8112106
Encompass_Preferred
Volumetric Flask, Class A, To Contain, 10mL +/- 0.02mL capacity. Stopper Size: 9. Calibrated to contain at 20degreeC. Flask has serial number and is supplied with a Certificate of Compliance.
Catalog No. 50-807-321
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.