Learn More
Diagnostic Biosystems FLT-1 Flt-1/VEGFR1
Supplier:  Diagnostic Biosystems RP077
Encompass_Preferred
This antibody reacts with a 180 kDa protein, known as Flt-1 or vascular endothelial growth factor receptor 1 (VEGFR1). Flt-1 is a cell membrane receptor kinase. Flk-1 has a high affinity receptor for vascular endothelial growth factor and is putatively involved in the growth of endothelial cells and angiogenesis.
Catalog No. 50-238-5428
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.